MAT089755866 Matrix Scientific

2-(1-Methyl-1H-benzoimidazol-2-yl)-ethylamine

  • MDL number MFCD01836868
  • Cas Number : 184959-13-5
  • Molecular Weight : 175.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018685.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018685.html
Availability details for
Quantity
  • Availability
  • Add to Cart