MAT089755882 Matrix Scientific

4-(4-Methyl-piperidine-1-sulfonyl)-phenylamine

  • MDL number MFCD02020755
  • Cas Number : 314285-39-7
  • Molecular Weight : 254.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018701.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018701.html
Availability details for
Quantity
  • Availability
  • Add to Cart