MAT089755887 Matrix Scientific

3-Chloro-2-piperidin-1-yl-phenylamine

  • MDL number MFCD04971011
  • Cas Number : 58785-06-1
  • Molecular Weight : 210.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018706.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018706.html
Availability details for
Quantity
  • Availability
  • Add to Cart