MAT089755931 Matrix Scientific

4-Methoxy-1H-indole-2-carboxylic acid

  • MDL number MFCD02664458
  • Cas Number : 103260-65-7
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018750.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018750.html
Availability details for
Quantity
  • Availability
  • Add to Cart