MAT089755946 Matrix Scientific

5-Methyl-2-(1-thiophen-2-yl-ethyl)-2H-pyrazol-3-ylamine

  • MDL number MFCD07186342
  • Cas Number : 957483-06-6
  • Molecular Weight : 207.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018765.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018765.html
Availability details for
Quantity
  • Availability
  • Add to Cart