MAT089755976 Matrix Scientific

Phenethyl-(1,2,5-trimethyl-piperidin-4-yl)-amine

  • MDL number MFCD04238113
  • Cas Number : 100096-38-6
  • Molecular Weight : 246.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 149-151�Torr
Product Link http://www.matrixscientific.com/018796.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018796.html
Availability details for
Quantity
  • Availability
  • Add to Cart