MAT089755977 Matrix Scientific

(1-Methyl-piperidin-4-yl)-(1-phenyl-ethyl)-amine

  • MDL number MFCD03640279
  • Cas Number : 359880-52-7
  • Molecular Weight : 218.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018797.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018797.html
Availability details for
Quantity
  • Availability
  • Add to Cart