MAT089755983 Matrix Scientific

(3-Amino-4-pyrrolidin-1-yl-phenyl)-pyrrolidin-1-yl-methanone

  • MDL number MFCD03928545
  • Cas Number : 640263-69-0
  • Molecular Weight : 259.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018803.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018803.html
Availability details for
Quantity
  • Availability
  • Add to Cart