MAT089755990 Matrix Scientific

4-Furan-2-yl-2,3,3a,4,5,9b-hexahydro-furo[3,2-c]-quinoline

  • MDL number MFCD06589803
  • Cas Number : 1164528-57-7
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018810.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018810.html
Availability details for
Quantity
  • Availability
  • Add to Cart