MAT089755997 Matrix Scientific

4-Phenyl-2,3,3a,4,5,9b-hexahydro-furo[3,2-c]-quinoline

  • MDL number MFCD03662167
  • Cas Number : 745787-25-1
  • Molecular Weight : 251.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018817.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018817.html
Availability details for
Quantity
  • Availability
  • Add to Cart