MAT089756011 Matrix Scientific

1-(3-Methoxy-phenyl)-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD04217650
  • Cas Number : 700854-99-5
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018831.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018831.html
Availability details for
Quantity
  • Availability
  • Add to Cart