MAT089756032 Matrix Scientific

2-Methyl-1-prop-2-ynyl-1H-indole-3-carbaldehyde

  • MDL number MFCD04342457
  • Cas Number : 842973-82-4
  • Molecular Weight : 197.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018853.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018853.html
Availability details for
Quantity
  • Availability
  • Add to Cart