MAT089756045 Matrix Scientific

2,5-Dimethyl-1-pyridin-2-yl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD04218024
  • Cas Number : 32570-90-4
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018866.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018866.html
Availability details for
Quantity
  • Availability
  • Add to Cart