MAT089756063 Matrix Scientific

1-Ethyl-2,3-dimethyl-1H-indole-5-carboxylic acid

  • MDL number MFCD02090858
  • Cas Number : 438218-59-8
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018886.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018886.html
Availability details for
Quantity
  • Availability
  • Add to Cart