MAT089756083 Matrix Scientific

2-Morpholin-4-yl-isonicotinic acid

  • MDL number MFCD03411666
  • Cas Number : 295349-64-3
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018906.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018906.html
Availability details for
Quantity
  • Availability
  • Add to Cart