MAT089756095 Matrix Scientific

5-(3,4-Dichloro-phenyl)-isoxazole-3-carboxylic acid

  • MDL number MFCD07186353
  • Cas Number : 876710-49-5
  • Molecular Weight : 258.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018918.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018918.html
Availability details for
Quantity
  • Availability
  • Add to Cart