MAT089756139 Matrix Scientific

Methyl-(3-piperidin-1-yl-propyl)-amine

  • MDL number MFCD07186366
  • Cas Number : 86010-41-5
  • Molecular Weight : 156.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018963.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018963.html
Availability details for
Quantity
  • Availability
  • Add to Cart