MAT089756183 Matrix Scientific

4-[2-(5-Methyl-1H-benzoimidazol-2-yl)-ethyl]-phenylamine

  • MDL number MFCD06067715
  • Cas Number : 883541-99-9
  • Molecular Weight : 251.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019007.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019007.html
Availability details for
Quantity
  • Availability
  • Add to Cart