MAT089756185 Matrix Scientific

(2-Methyl-piperidin-1-yl)-acetic acid

  • MDL number MFCD06589812
  • Cas Number : 856437-61-1
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019009.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019009.html
Availability details for
Quantity
  • Availability
  • Add to Cart