MAT089756214 Matrix Scientific

1-(3-Ethoxy-propyl)-5-oxo-pyrrolidine-3-carboxylic acid

  • MDL number MFCD04989730
  • Cas Number : 407634-06-4
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019038.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019038.html
Availability details for
Quantity
  • Availability
  • Add to Cart