MAT089756215 Matrix Scientific

1,1-Dimethyl-2-(4-methyl-piperidin-1-yl)-ethylamine

  • MDL number MFCD05022424
  • Cas Number : 690632-11-2
  • Molecular Weight : 170.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019039.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019039.html
Availability details for
Quantity
  • Availability
  • Add to Cart