MAT089756218 Matrix Scientific

3-Azepan-1-yl-2,2-dimethyl-propionaldehyde

  • MDL number MFCD05022485
  • Cas Number : 842971-09-9
  • Molecular Weight : 183.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019042.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019042.html
Availability details for
Quantity
  • Availability
  • Add to Cart