MAT089756221 Matrix Scientific

2,3-Diethyl-quinoxaline-6-carboxylic acid

  • MDL number MFCD03450169
  • Cas Number : 694506-87-1
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019045.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019045.html
Availability details for
Quantity
  • Availability
  • Add to Cart