MAT089756243 Matrix Scientific

3-(3-m-Tolyl-[1,2,4]oxadiazol-5-yl)-propionic acid

  • MDL number MFCD07186380
  • Cas Number : 94192-16-2
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019067.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019067.html
Availability details for
Quantity
  • Availability
  • Add to Cart