MAT089756260 Matrix Scientific

2-(2-Methyl-2H-pyrazol-3-yl)-quinoline-4-carboxylic acid

  • MDL number MFCD04969978
  • Cas Number : 380489-22-5
  • Molecular Weight : 253.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019084.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019084.html
Availability details for
Quantity
  • Availability
  • Add to Cart