MAT089756270 Matrix Scientific

2,6-Dimethyl-1H-indole-3-carbaldehyde

  • MDL number MFCD06016187
  • Cas Number : 728024-59-7
  • Molecular Weight : 173.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019094.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019094.html
Availability details for
Quantity
  • Availability
  • Add to Cart