MAT089756343 Matrix Scientific

(4-Pyrrol-1-yl-phenoxy)-acetic acid

  • MDL number MFCD05258773
  • Cas Number : 404892-58-6
  • Molecular Weight : 217.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019167.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019167.html
Availability details for
Quantity
  • Availability
  • Add to Cart