MAT089756349 Matrix Scientific

(1-Isopropyl-piperidin-3-ylmethyl)-methyl-amine

  • MDL number MFCD07643226
  • Cas Number : 876716-01-7
  • Molecular Weight : 170.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019173.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019173.html
Availability details for
Quantity
  • Availability
  • Add to Cart