MAT089756366 Matrix Scientific

4-Methyl-2-tetrazol-1-yl-pentanoic acid

  • MDL number MFCD07643230
  • Cas Number : 876716-26-6
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019190.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019190.html
Availability details for
Quantity
  • Availability
  • Add to Cart