MAT089756372 Matrix Scientific

(4-Amino-3,5-dimethyl-pyrazol-1-yl)-acetic acid

  • MDL number MFCD04969595
  • Cas Number : 956354-80-6
  • Molecular Weight : 169.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019196.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019196.html
Availability details for
Quantity
  • Availability
  • Add to Cart