MAT089756383 Matrix Scientific

5-(4-Methoxy-phenyl)-1-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD07186421
  • Cas Number : 957312-76-4
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019207.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019207.html
Availability details for
Quantity
  • Availability
  • Add to Cart