MAT089756412 Matrix Scientific

2-Dimethylamino-isonicotinic acid

  • MDL number MFCD07643262
  • Cas Number : 77314-81-9
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.247
Flash Point 204
Country of Origin USA
Product Link http://www.matrixscientific.com/019238.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019238.html
Availability details for
Quantity
  • Availability
  • Add to Cart