MAT089756428 Matrix Scientific

1,1-Dimethyl-2,3,4,9-tetrahydro-1H-beta-carboline-3-carboxylic acid

  • MDL number MFCD00277074
  • Cas Number : 73198-03-5
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019254.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019254.html
Availability details for
Quantity
  • Availability
  • Add to Cart