MAT089756438 Matrix Scientific

3-tert-Butyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD07186448
  • Cas Number : 1001020-17-2
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019264.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019264.html
Availability details for
Quantity
  • Availability
  • Add to Cart