MAT089756452 Matrix Scientific

5-Chloro-3-methyl-1H-indole-2-carboxylic acid

  • MDL number MFCD05864560
  • Cas Number : 16381-47-8
  • Molecular Weight : 209.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019278.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019278.html
Availability details for
Quantity
  • Availability
  • Add to Cart