MAT089756497 Matrix Scientific

1-Piperazin-1-yl-3-p-tolyloxy-propan-2-ol

  • MDL number MFCD07186460
  • Cas Number : 883546-08-5
  • Molecular Weight : 250.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019323.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019323.html
Availability details for
Quantity
  • Availability
  • Add to Cart