MAT089756616 Matrix Scientific

4-(1H-Benzoimidazol-2-yl)-2-methyl-phenylamine

  • MDL number MFCD05843142
  • Cas Number : 874591-54-5
  • Molecular Weight : 223.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019431.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019431.html
Availability details for
Quantity
  • Availability
  • Add to Cart