MAT089756623 Matrix Scientific

4,5,6-Trimethoxy-1-methyl-1H-indole-2-carboxylic acid

  • MDL number MFCD03848073
  • Cas Number : 481074-71-9
  • Molecular Weight : 265.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019438.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019438.html
Availability details for
Quantity
  • Availability
  • Add to Cart