MAT089756653 Matrix Scientific

(1-Cyclopentyl-3-oxo-piperazin-2-yl)-acetic acid

  • MDL number MFCD05666764
  • Cas Number : 1042697-00-6
  • Molecular Weight : 226.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019468.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019468.html
Availability details for
Quantity
  • Availability
  • Add to Cart