MAT089756664 Matrix Scientific

3-(4-Sulfamoyl-phenyl)-propionic acid

  • MDL number MFCD06659895
  • Cas Number : 90610-69-8
  • Molecular Weight : 229.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-150
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/019479.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019479.html
Availability details for
Quantity
  • Availability
  • Add to Cart