MAT089756696 Matrix Scientific

(3-Formyl-2-methyl-indol-1-yl)-acetic acid

  • MDL number MFCD02909101
  • Cas Number : 432001-45-1
  • Molecular Weight : 217.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019511.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019511.html
Availability details for
Quantity
  • Availability
  • Add to Cart