MAT089756756 Matrix Scientific

3-(3-Methyl-piperidin-1-yl)-propionic acid

  • MDL number MFCD06753040
  • Cas Number : 858444-38-9
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019571.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019571.html
Availability details for
Quantity
  • Availability
  • Add to Cart