MAT089756791 Matrix Scientific

1-(4-Methoxy-benzyl)-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD07396162
  • Cas Number : 861162-64-3
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/019607.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019607.html
Availability details for
Quantity
  • Availability
  • Add to Cart