MAT089756797 Matrix Scientific

3-Furan-2-yl-4-methyl-pentanoic acid

  • MDL number MFCD07186513
  • Cas Number : 842954-66-9
  • Molecular Weight : 182.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019613.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019613.html
Availability details for
Quantity
  • Availability
  • Add to Cart