MAT089756806 Matrix Scientific

2-Cyclopentyl-3-oxo-2,3-dihydro-1H-isoindole-4-carboxylic acid

  • MDL number MFCD06653224
  • Cas Number : 878424-59-0
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019622.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019622.html
Availability details for
Quantity
  • Availability
  • Add to Cart