MAT089756816 Matrix Scientific

5-(4-Methoxy-phenyl)-4-phenyl-2,4-dihydro-[1,2,4]triazole-3-thione

  • MDL number MFCD01151900
  • Cas Number : 91759-68-1
  • Molecular Weight : 283.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 284-285 �ec)
Country of Origin USA
Product Link http://www.matrixscientific.com/019632.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019632.html
Availability details for
Quantity
  • Availability
  • Add to Cart