MAT089756850 Matrix Scientific

2-Pyrrolidin-2-yl-1H-benzoimidazole

  • MDL number MFCD01859287
  • Cas Number : 638141-64-7
  • Molecular Weight : 187.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019666.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019666.html
Availability details for
Quantity
  • Availability
  • Add to Cart