MAT089756853 Matrix Scientific

2-(3,5-Dimethyl-1H-pyrazol-4-yl)-ethylamine

  • MDL number MFCD03043480
  • Cas Number : 423176-38-9
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019669.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019669.html
Availability details for
Quantity
  • Availability
  • Add to Cart