MAT089756873 Matrix Scientific

4-Methyl-3-(1H-pyrrol-1-yl)aniline

  • MDL number MFCD07643299
  • Cas Number : 94009-17-3
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019689.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019689.html
Availability details for
Quantity
  • Availability
  • Add to Cart