MAT089756876 Matrix Scientific

3,5-Dimethyl-1-(2-methyl-allyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD07186516
  • Cas Number : 1004365-13-2
  • Molecular Weight : 178.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019692.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019692.html
Availability details for
Quantity
  • Availability
  • Add to Cart